accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 541-88-8

IUPAC Name: 2-chloroacetyl 2-chloroacetate
Synonyms: chloroacetic anhydride
Molecular Weight (g/mol): 170.97
Molecular Formula: C4H4Cl2O3
InChi Key: PNVPNXKRAUBJGW-UHFFFAOYSA-N
SMILES: ClCC(=O)OC(=O)CCl
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)