accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOBrBr

CAS RN 54113-41-6

IUPAC Name: methyl 4,5-dibromofuran-2-carboxylate
Synonyms: methyl 4,5-dibromofuran-2-carboxylate
Molecular Weight (g/mol): 283.9
Molecular Formula: C6H4Br2O3
InChi Key: OIOPUTCGINQNRL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=C(Br)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results