accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClClCl

CAS RN 54135-80-7

IUPAC Name: 1,2,3-trichloro-4-methoxybenzene
Synonyms: 1,2,3-trichloro-4-methoxybenzene
Molecular Weight (g/mol): 211.47
Molecular Formula: C7H5Cl3O
InChi Key: FRQUNVLMWIYOLV-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results