accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrN

CAS RN 5414-21-1

IUPAC Name: 5-bromopentanenitrile
Synonyms: 5-bromopentanenitrile
Molecular Weight (g/mol): 162.03
Molecular Formula: C5H8BrN
InChi Key: NWWWGAKVHCSAEU-UHFFFAOYSA-N
SMILES: BrCCCCC#N
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)