accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHBrBr

CAS RN 541545-55-5

IUPAC Name: N-(2,4-dibromophenyl)-4-methoxybenzamide
Synonyms: N-(2,4-dibromophenyl)-4-methoxybenzamide
Molecular Weight (g/mol): 385.06
Molecular Formula: C14H11Br2NO2
InChi Key: MEEOWNOOARIQDA-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NC1=C(Br)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results