accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNO

CAS RN 5416-80-8

IUPAC Name: 2-methyl-1H-indole-3-carbaldehyde
Synonyms: 2-methyl-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: CYZIVXOEJNAIBS-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)