accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5417-17-4

IUPAC Name: 2-chloro-3,4-dimethoxybenzaldehyde
Synonyms: 2-chloro-3,4-dimethoxybenzaldehyde
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: SAWHDJTZESXNMM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(Cl)=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results