accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHO

CAS RN 5417-63-0

IUPAC Name: 3-aminonaphthalen-2-ol
Synonyms: 3-aminonaphthalen-2-ol
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: ZHVPTERSBUMMHK-UHFFFAOYSA-N
SMILES: NC1=C(O)C=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)