accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3NN

CAS RN 5417-82-3

IUPAC Name: 2-(1-ethoxyethylidene)propanedinitrile
Synonyms: 2-(1-ethoxyethylidene)propanedinitrile
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: BOSVWXDDFBSSIZ-UHFFFAOYSA-N
SMILES: CCOC(C)=C(C#N)C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)