accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5418-32-6

IUPAC Name: sodium 4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Synonyms: sodium 4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzenolate
Molecular Weight (g/mol): 221.19
Molecular Formula: C12H8NNaO2
InChi Key: FRUIYGAEJNTDCA-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC=C(C=C1)N=C1C=CC(=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results