accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNNClClCl

CAS RN 54185-42-1

IUPAC Name: 2,4,6,8-tetrachloroquinazoline
Synonyms: 2,4,6,8-tetrachloroquinazoline
Molecular Weight (g/mol): 267.92
Molecular Formula: C8H2Cl4N2
InChi Key: GYUFSVVBAINEBI-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C2N=C(Cl)N=C(Cl)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results