accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-08-5

IUPAC Name: propan-2-yl 3-oxobutanoate
Synonyms: isopropyl 3-oxobutanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: GVIIRWAJDFKJMJ-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CC(C)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results