accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-18-7

IUPAC Name: chlorocyclohexane
Synonyms: chlorocyclohexane C6H11cl chlorzyklohexan
Molecular Weight (g/mol): 118.6
Molecular Formula: C6H11Cl
InChi Key: UNFUYWDGSFDHCW-UHFFFAOYSA-N
SMILES: ClC1CCCCC1
Boiling Point 
  • (2)
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)