accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-32-5

IUPAC Name: 2-aminohexanedioic acid
Synonyms: aminoadipic acid Aad
Molecular Weight (g/mol): 161.16
Molecular Formula: C6H11NO4
InChi Key: OYIFNHCXNCRBQI-UHFFFAOYNA-N
SMILES: NC(CCCC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results