accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3N

CAS RN 542-54-1

IUPAC Name: 4-methylpentanenitrile
Synonyms: 4-methylpentanitrile
Molecular Weight (g/mol): 97.16
Molecular Formula: C6H11N
InChi Key: DUJMVKJJUANUMQ-UHFFFAOYSA-N
SMILES: CC(C)CCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results