accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-56-3

IUPAC Name: 2-methylpropyl nitrite
Synonyms: isobutyl nitrite isobutylnitrit isobutyl nitrite (substance) IBN blackjack
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: APNSGVMLAYLYCT-UHFFFAOYSA-N
SMILES: CC(C)CON=O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)