accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-69-8

IUPAC Name: 1-iodobutane
Synonyms: butyl iodide N-butyl iodide
Molecular Weight (g/mol): 184.02
Molecular Formula: C4H9I
InChi Key: KMGBZBJJOKUPIA-UHFFFAOYSA-N
SMILES: CCCCI
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)