accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-90-5

IUPAC Name: (ethylsulfanyl)carbonitrile
Synonyms: ethyl thiocyanate
Molecular Weight (g/mol): 87.14
Molecular Formula: C3H5NS
InChi Key: WFCLYEAZTHWNEH-UHFFFAOYSA-N
SMILES: CCSC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results