accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONO

CAS RN 54221-96-4

IUPAC Name: 6-methoxypyridine-2-carbaldehyde
Synonyms: 6-methoxypyridine-2-carbaldehyde
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: YDNWTNODZDSPNZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=N1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)