accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 5424-19-1

IUPAC Name: 3-benzoylpyridine
Synonyms: phenyl-B-pyridyl ketone
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: RYMBAPVTUHZCNF-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=CN=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)