accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2HOOHO

CAS RN 5424-29-3

IUPAC Name: 2-amino-3-hydroxy-2-methylpropanoic acid
Synonyms: 2-methylserine
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: CDUUKBXTEOFITR-UHFFFAOYNA-N
SMILES: CC(N)(CO)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)