accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54256-43-8

IUPAC Name: 4-decylbenzoyl chloride
Synonyms: 4-decylbenzoyl chloride
Molecular Weight (g/mol): 280.84
Molecular Formula: C17H25ClO
InChi Key: HEWLDHSFOQXCSI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=C(C=C1)C(Cl)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)