accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOH3CBr

CAS RN 5426-65-3

IUPAC Name: 4-bromo-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Synonyms: 4-bromo-1,5-dimethyl-2-phenylpyrazol-3-one
Molecular Weight (g/mol): 267.13
Molecular Formula: C11H11BrN2O
InChi Key: QYWWSDVPNCBSIA-UHFFFAOYSA-N
SMILES: CN1N(C(=O)C(Br)=C1C)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results