CAS RN 5428-09-1
IUPAC Name:
2-(prop-2-en-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms:
2-(prop-2-en-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol):
187.2
Molecular Formula:
C11H9NO2
InChi Key:
MHHGQWMCVNQHLG-UHFFFAOYSA-N
SMILES:
C=CCN1C(=O)C2=CC=CC=C2C1=O
1
–
2
of
2
results
1
N-Allylphthalimide 98.0+%, TCI America™
CAS: 5428-09-1 Molecular Formula: C11H9NO2 Molecular Weight (g/mol): 187.20 MDL Number: MFCD00158662 InChI Key: MHHGQWMCVNQHLG-UHFFFAOYSA-N Synonym: 2-allylisoindoline-1,3-dione,n-allylphthalimide,n-2-propenyl phthalimide,2-allyl-1h-isoindole-1,3 2h-dione,2-prop-2-en-1-yl-1h-isoindole-1,3 2h-dione,1h-isoindole-1,3 2h-dione, 2-2-propen-1-yl,2-prop-2-en-1-yl-2,3-dihydro-1h-isoindole-1,3-dione,n-allylphthalimid,n-allyl-phthalimide,2-allylisoindole-1,3-dione PubChem CID: 224401 IUPAC Name: 2-prop-2-enylisoindole-1,3-dione SMILES: C=CCN1C(=O)C2=CC=CC=C2C1=O
2
N-Allylphthalimide, 97%, Thermo Scientific™
CAS: 5428-09-1 MDL Number: MFCD00158662