accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OClO

CAS RN 543-27-1

IUPAC Name: 2-methylpropyl carbonochloridate
Synonyms: 2-methylpropyl carbonochloridate
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: YOETUEMZNOLGDB-UHFFFAOYSA-N
SMILES: CC(C)COC(Cl)=O
Boiling Point 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)