accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5431-44-7

IUPAC Name: pyridine-2,6-dicarbaldehyde
Synonyms: pyridine-2,6-dicarbaldehyde
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: PMWXGSWIOOVHEQ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(C=O)=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)