accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONSHOO

CAS RN 54323-50-1

IUPAC Name: 3-acetyl-1,3-thiazolidine-4-carboxylic acid
Synonyms: 3-acetyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol): 175.2
Molecular Formula: C6H9NO3S
InChi Key: WXTBYSIPOKXCPM-UHFFFAOYNA-N
SMILES: CC(=O)N1CSCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results