accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONHO

CAS RN 5435-54-1

IUPAC Name: 3-nitro-1,4-dihydropyridin-4-one
Synonyms: 3-nitro-1H-pyridin-4-one
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: YUWOLBZMQDGRFV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC=CC1=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)