accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5436-01-1

IUPAC Name: 1-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione
Synonyms: 1-methyl-2H-pyridazine-3,6-dione
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: UAECOHJYXUJDOF-UHFFFAOYSA-N
SMILES: CN1NC(=O)C=CC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results