accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5437-98-9

IUPAC Name: N-(4-methoxyphenyl)-3-oxobutanamide
Synonyms: N-(4-methoxyphenyl)-3-oxobutanamide
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: SWAJJKROCOJICG-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)CC(C)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results