accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 543717-62-0

IUPAC Name: 4-chloro-N-propylbutanamide
Synonyms: 4-chloro-N-propylbutanamide
Molecular Weight (g/mol): 163.65
Molecular Formula: C7H14ClNO
InChi Key: JWDRIUIZEWDSCP-UHFFFAOYSA-N
SMILES: CCCNC(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results