accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 544-13-8

IUPAC Name: pentanedinitrile
Synonyms: glutaronitrile
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: ZTOMUSMDRMJOTH-UHFFFAOYSA-N
SMILES: N#CCCCC#N
Boiling Point 
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)