accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 544-25-2

IUPAC Name: cyclohepta-1,3,5-triene
Synonyms: cycloheptatriene tropilidene tropiliden tropilidine tropyliden 1H-[7]annulene zykloheptatrien cycloheptatrien
Molecular Weight (g/mol): 92.14
Molecular Formula: C7H8
InChi Key: CHVJITGCYZJHLR-UHFFFAOYSA-N
SMILES: C1C=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)