accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 5441-51-0

IUPAC Name: 4-ethylcyclohexan-1-one
Synonyms: cyclohexanone, 4-ethyl-
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: OKSDJGWHKXFVME-UHFFFAOYSA-N
SMILES: CCC1CCC(=O)CC1
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)