accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5441-52-1

IUPAC Name: 3,5-dimethylcyclohexan-1-ol
Synonyms: 3,5-dimethylcyclohexanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: WIYNOPYNRFPWNB-UHFFFAOYNA-N
SMILES: CC1CC(C)CC(O)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results