accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5442-24-0

IUPAC Name: 6-amino-5-nitroso-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-amino-5-nitroso-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 156.1
Molecular Formula: C4H4N4O3
InChi Key: DKPCSXFEWFSECE-UHFFFAOYSA-N
SMILES: NC1=C(N=O)C(=O)NC(=O)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results