accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5443-49-2

IUPAC Name: (2E)-2-bromo-3-phenylprop-2-enal
Synonyms: bromocinnamal
Molecular Weight (g/mol): 211.06
Molecular Formula: C9H7BrO
InChi Key: WQRWNOKNRHCLHV-RMKNXTFCSA-N
SMILES: Br\C(C=O)=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)