accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NN

CAS RN 5444-01-9

IUPAC Name: 4-methylpyridine-3-carbonitrile
Synonyms: 4-methylpyridine-3-carbonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: XLAPHZHNODDMDD-UHFFFAOYSA-N
SMILES: CC1=CC=NC=C1C#N
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)