accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNBrBr

CAS RN 544436-46-6

IUPAC Name: 2,7-dibromo-9-(2-ethylhexyl)-9H-carbazole
Synonyms: 2,7-dibromo-9-(2-ethylhexyl)carbazole
Molecular Weight (g/mol): 437.22
Molecular Formula: C20H23Br2N
InChi Key: OLOMDFPWMJQODN-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results