accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 544478-19-5

H3COONNOOOOH3CCH3CHCH3

CAS RN 544478-19-5

IUPAC Name: 3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1-(prop-2-yn-1-yl)-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: 3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1-(prop-2-yn-1-yl)-4H-pyridine-3,5-dicarboxylate
Molecular Weight (g/mol): 398.42
Molecular Formula: C21H22N2O6
InChi Key: BITHABUTZRAUGT-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C(C)N(CC#C)C(C)=C(C1C1=CC(=CC=C1)[N+]([O-])=O)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results