CAS RN 5445-24-9
CAS RN 5445-24-9
IUPAC Name:
1-propylcyclohexan-1-ol
Synonyms:
1-propylcyclohexan-1-ol
Molecular Weight (g/mol):
142.24
Molecular Formula:
C9H18O
InChi Key:
PYLPYOPJKOJRNP-UHFFFAOYSA-N
SMILES:
CCCC1(O)CCCCC1
1
–
1
of
1
results
1
1-n-Propylcyclohexanol, 98%, Thermo Scientific Chemicals
CAS: 5445-24-9 Molecular Formula: C9H18O Molecular Weight (g/mol): 142.24 MDL Number: MFCD00046364 InChI Key: PYLPYOPJKOJRNP-UHFFFAOYSA-N Synonym: 1-propylcyclohexanol,1-n-propylcyclohexanol,cyclohexanol, 1-propyl,1-n-propyl cyclohexanol-1,propylcyclohexanol,1-n-propyl-1-cyclohexanol,1-n-propylcyclohexane-1-ol PubChem CID: 138504 IUPAC Name: 1-propylcyclohexan-1-ol SMILES: CCCC1(O)CCCCC1