accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54451-24-0

IUPAC Name: dilanthanum(3+) tricarbonate
Synonyms: lanthanum carbonate
Molecular Weight (g/mol): 457.84
Molecular Formula: C3La2O9
InChi Key: NZPIUJUFIFZSPW-UHFFFAOYSA-H
SMILES: [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)