accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5446-05-9

ONOClI

CAS RN 5446-05-9

IUPAC Name: 4-chloro-1-iodo-2-nitrobenzene
Synonyms: 4-chloro-1-iodo-2-nitrobenzene
Molecular Weight (g/mol): 283.45
Molecular Formula: C6H3ClINO2
InChi Key: ZLQFWQOAQQBLLZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CC=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results