accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 5447-02-9

IUPAC Name: 3,4-bis(benzyloxy)benzaldehyde
Synonyms: 3,4-bis(benzyloxy)benzaldehyde
Molecular Weight (g/mol): 318.37
Molecular Formula: C21H18O3
InChi Key: XDDLXZHBWVFPRG-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)