accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CBrNOO

CAS RN 5447-97-2

IUPAC Name: 2-bromo-2-nitropropane
Synonyms: 2-bromo-2-nitro-propane
Molecular Weight (g/mol): 167.99
Molecular Formula: C3H6BrNO2
InChi Key: OADSZWXMXIWZSQ-UHFFFAOYSA-N
SMILES: CC(C)(Br)[N+]([O-])=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)