accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5448-36-2

IUPAC Name: 3-methyl-N,N-bis(propan-2-yl)benzamide
Synonyms: N,N-diisopropyl-3-methylbenzamide
Molecular Weight (g/mol): 219.33
Molecular Formula: C14H21NO
InChi Key: WPFBQNDWEOVSGC-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)C1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results