accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 5452-75-5

IUPAC Name: ethyl 2-cyclohexylacetate
Synonyms: ethyl cyclohexylacetate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: ZBDAMDWKXGTKBT-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results