accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54523-47-6

IUPAC Name: 1,3-bis(4-bromophenyl)propan-1-one
Synonyms: 1,3-bis(4-bromophenyl)propan-1-one
Molecular Weight (g/mol): 368.07
Molecular Formula: C15H12Br2O
InChi Key: PQDQAUVBWSUMMB-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CCC(=O)C2=CC=C(Br)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results