accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 5453-80-5

IUPAC Name: bicyclo[2.2.1]hept-5-ene-2-carbaldehyde
Synonyms: bicyclo[2.2.1]hept-5-ene-2-carbaldehyde
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: AJIBZRIAUXVGQJ-UHFFFAOYNA-N
SMILES: O=CC1CC2CC1C=C2
Boiling Point 
  • (1)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)