accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 545394-83-0

IUPAC Name: 2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde
Synonyms: 2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 189.14
Molecular Formula: C8H6F3NO
InChi Key: HHGVNSFXEFHMGQ-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=CC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results